Learn More
Biotium Primary Antibody CD4 SK3 PerCP 1/EA
Catalog No. 5000498364
Encompass_Preferred
CD4 (clone SK3) is a mouse monoclonal antibody that recognizes the human protein CD4 CD4 is a common marker of Helper T cells and is a T cell coreceptor This PerCP antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-98364 Supplier Biotium Supplier No. P001PCP500
CD4 (clone SK3) is a mouse monoclonal antibody that recognizes the human protein CD4 CD4 is a common marker of Helper T cells and is a T cell coreceptor This PerCP antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests