Learn More
Biotium Primary Antibody CD45 2D1 PerCP 1/EA
Catalog No. 5000497614
Encompass_Preferred
CD45 (clone 2D1) is a mouse monoclonal antibody that recognizes the human protein CD45 CD45 is also known as Leukocyte Common Antigen (LCA) This PerCP antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-97614 Supplier Biotium Supplier No. P012PCP500
CD45 (clone 2D1) is a mouse monoclonal antibody that recognizes the human protein CD45 CD45 is also known as Leukocyte Common Antigen (LCA) This PerCP antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests