Learn More
Biotium Primary Antibody CD45 2D1 RPE Astral775 1/EA
Catalog No. 5000497189
Encompass_Preferred
CD45 (clone 2D1) is a mouse monoclonal antibody that recognizes the human protein CD45 CD45 is also known as Leukocyte Common Antigen (LCA) This RPE-Astral775 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-97189 Supplier Biotium Supplier No. P012R775125
CD45 (clone 2D1) is a mouse monoclonal antibody that recognizes the human protein CD45 CD45 is also known as Leukocyte Common Antigen (LCA) This RPE-Astral775 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests