Learn More
Biotium Primary Antibody CD56 r5.1H11 CF488A 1/EA
Catalog No. 5000500443
Encompass_Preferred
CD56 (r5 1 H11) is a recombinant mouse monoclonal antibody that recognizes the human protein CD56 This CF488A antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-005-00443 Supplier Biotium Supplier No. P026488A500
CD56 (r5 1 H11) is a recombinant mouse monoclonal antibody that recognizes the human protein CD56 This CF488A antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests