Learn More
Biotium Primary Antibody CD56 r5.1H11 CF647Plus 1/EA
Catalog No. 5000499967
Encompass_Preferred
CD56 (r5 1 H11) is a recombinant mouse monoclonal antibody that recognizes the human protein CD56 This CF647Plus antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-99967 Supplier Biotium Supplier No. P026647P125
CD56 (r5 1 H11) is a recombinant mouse monoclonal antibody that recognizes the human protein CD56 This CF647Plus antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests