Learn More
Biotium Primary Antibody CD56 r5.1H11 CF820 1/EA
Catalog No. 5000499458
Encompass_Preferred
CD56 (r5 1 H11) is a recombinant mouse monoclonal antibody that recognizes the human protein CD56 This CF820 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-99458 Supplier Biotium Supplier No. P026820500
CD56 (r5 1 H11) is a recombinant mouse monoclonal antibody that recognizes the human protein CD56 This CF820 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests