Learn More
Biotium Primary Antibody CD56 r5.1H11 R PE 1/EA
Catalog No. 5000499668
Encompass_Preferred
CD56 (r5 1 H11) is a recombinant mouse monoclonal antibody that recognizes the human protein CD56 This RPE antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-99668 Supplier Biotium Supplier No. P026RPE125
CD56 (r5 1 H11) is a recombinant mouse monoclonal antibody that recognizes the human protein CD56 This RPE antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests