Learn More
Biotium Primary Antibody CD63 2304.63 CF488A 1/EA
Catalog No. 5000497532
Encompass_Preferred
CD63 (2304 63) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD63 This CF488A antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-97532 Supplier Biotium Supplier No. P013488A125
CD63 (2304 63) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD63 This CF488A antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests