Learn More
Biotium Primary Antibody CD63 2304.63 CF568 1/EA
Catalog No. 5000497942
Encompass_Preferred
CD63 (2304 63) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD63 This CF568 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-97942 Supplier Biotium Supplier No. P013568500
Add to Cart
Add to Cart
CD63 (2304 63) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD63 This CF568 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests