Learn More
Biotium Primary Antibody CD63 2304.63 CF583R 1/EA
Catalog No. 5000497542
Encompass_Preferred
CD63 (2304 63) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD63 This CF583R antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-97542 Supplier Biotium Supplier No. P013583R500
CD63 (2304 63) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD63 This CF583R antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests