Learn More
Biotium Primary Antibody CD63 2304.63 RPE Astral775 1/EA
Catalog No. 5000497196
Encompass_Preferred
CD63 (2304 63) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD63 This RPE-Astral775 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-97196 Supplier Biotium Supplier No. P013R775500
CD63 (2304 63) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD63 This RPE-Astral775 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests