Learn More
Biotium Primary Antibody CD63 24MS05.63 CF405M 1/EA
Catalog No. 5000499126
Encompass_Preferred
CD63 (24MS05 63) is a recombinant rat monoclonal antibody that recognizes the mouse tetraspanin protein CD63 This CF405M antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-99126 Supplier Biotium Supplier No. P020405M500
Add to Cart
Add to Cart
CD63 (24MS05 63) is a recombinant rat monoclonal antibody that recognizes the mouse tetraspanin protein CD63 This CF405M antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests