Learn More
Biotium Primary Antibody CD63 24MS05.63 CF568 1/EA
Catalog No. 5000496777
Encompass_Preferred
CD63 (24MS05 63) is a recombinant rat monoclonal antibody that recognizes the mouse tetraspanin protein CD63 This CF568 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-96777 Supplier Biotium Supplier No. P020568500
CD63 (24MS05 63) is a recombinant rat monoclonal antibody that recognizes the mouse tetraspanin protein CD63 This CF568 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests