Learn More
Biotium Primary Antibody CD63 NA CF568 1/EA
Catalog No. 5000497623
Encompass_Preferred
ExoBrite STORM CF568 CD63 (Mouse) Antibody is designed for detection of extracellular vesicles by STORM CF568 has been validated and recommended for STORM microscopy and has Ex/Em at 562/583 nm 500 uL
Catalog No. 50-004-97623 Supplier Biotium Supplier No. P022568ST500
Add to Cart
Add to Cart
ExoBrite STORM CF568 CD63 (Mouse) Antibody is designed for detection of extracellular vesicles by STORM CF568 has been validated and recommended for STORM microscopy and has Ex/Em at 562/583 nm 500 uL