Learn More
Biotium Primary Antibody CD63 P08962 ExoBrite 640 660 1/EA
Catalog No. 5000497512
Encompass_Preferred
ExoBrite 640/660 CD63 Flow Antibody is optimized for detection of the exosome marker CD63 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 640/660 nm Recommended for instruments optimized for small particle flow 25 tests
Catalog No. 50-004-97512 Supplier Biotium Supplier No. P004640125
ExoBrite 640/660 CD63 Flow Antibody is optimized for detection of the exosome marker CD63 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 640/660 nm Recommended for instruments optimized for small particle flow 25 tests