Learn More
Biotium Primary Antibody CD81 2304.81 CF488A 1/EA
Catalog No. 5000497725
Encompass_Preferred
CD81 (2304 81) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD81 This CF488A antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-97725 Supplier Biotium Supplier No. P014488A500
Add to Cart
Add to Cart
CD81 (2304 81) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD81 This CF488A antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests