Learn More
Biotium Primary Antibody CD81 2304.81 RPE Astral616 1/EA
Catalog No. 5000497530
Encompass_Preferred
CD81 (2304 81) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD81 This RPE-Astral616 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-97530 Supplier Biotium Supplier No. P014R616500
CD81 (2304 81) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD81 This RPE-Astral616 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests