Learn More
Biotium Primary Antibody CD81 24MS04.81 CF488A 1/EA
Catalog No. 5000497599
Encompass_Preferred
CD81 (24MS04 81) is a recombinant hamster monoclonal antibody that recognizes the mouse and rat tetraspanin protein CD81 This CF488A antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-97599 Supplier Biotium Supplier No. P017488A500
Add to Cart
Add to Cart
CD81 (24MS04 81) is a recombinant hamster monoclonal antibody that recognizes the mouse and rat tetraspanin protein CD81 This CF488A antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests