Learn More
Biotium Primary Antibody CD81 NA CF647Plus 1/EA
Catalog No. 5000497364
Encompass_Preferred
ExoBrite STORM CF647Plus CD81 (Mouse/Rat) Antibody is designed for detection of extracellular vesicles by STORM CF647Plus has been validated and recommended for STORM microscopy and has Ex/Em at 652/668 nm 500 uL
Catalog No. 50-004-97364 Supplier Biotium Supplier No. P019647PST500
ExoBrite STORM CF647Plus CD81 (Mouse/Rat) Antibody is designed for detection of extracellular vesicles by STORM CF647Plus has been validated and recommended for STORM microscopy and has Ex/Em at 652/668 nm 500 uL