Learn More
Biotium Primary Antibody CD81 NA ExoBrite 490 515 1/EA
Catalog No. 5000497191
Encompass_Preferred
ExoBrite 490/515 CD81 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 490/515 nm for detection in the FITC channel Recommended for instruments optimized for small particle flow 100 tests
Catalog No. 50-004-97191 Supplier Biotium Supplier No. P019490500
ExoBrite 490/515 CD81 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 490/515 nm for detection in the FITC channel Recommended for instruments optimized for small particle flow 100 tests