Learn More
Biotium Primary Antibody CD81 NA ExoBrite 560 585 1/EA
Supplier:  Biotium P019560125
Encompass_Preferred
ExoBrite 560/585 CD81 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 562/583 nm for detection in the PE channel off the yellow laser line Recommended for instruments optimized for small particle flow 25 tests
Catalog No. 50-004-99072