Learn More
Biotium Primary Antibody CD81 NA ExoBrite 640 660 1/EA
Catalog No. 5000499073
Encompass_Preferred
ExoBrite 640/660 CD81 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 640/660 nm Recommended for instruments optimized for small particle flow 25 tests
Catalog No. 50-004-99073 Supplier Biotium Supplier No. P019640125
ExoBrite 640/660 CD81 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 640/660 nm Recommended for instruments optimized for small particle flow 25 tests