Learn More
Biotium Primary Antibody CD81 NA ExoBrite APC 1/EA
Catalog No. 5000499074
Encompass_Preferred
ExoBrite APC CD81 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry Recommended for instruments optimized for small particle flow 100 tests
Catalog No. 50-004-99074 Supplier Biotium Supplier No. P019APC500
ExoBrite APC CD81 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry Recommended for instruments optimized for small particle flow 100 tests