Learn More
Biotium Primary Antibody CD81 P60033 CF498 1/EA
Catalog No. 5000499104
Encompass_Preferred
ExoBrite STORM CF498 CD81 Antibody is designed for detection of extracellular vesicles by STORM CF498 has been validated and recommended for STORM microscopy and has Ex/Em at 498/518 nm 500 uL
Catalog No. 50-004-99104 Supplier Biotium Supplier No. P005498ST500
ExoBrite STORM CF498 CD81 Antibody is designed for detection of extracellular vesicles by STORM CF498 has been validated and recommended for STORM microscopy and has Ex/Em at 498/518 nm 500 uL