Learn More
Biotium Primary Antibody CD81 P60033 CF583R 1/EA
Catalog No. 5000497697
Encompass_Preferred
ExoBrite STORM CF583R CD81 Antibody is designed for detection of extracellular vesicles by STORM CF583R has been validated and recommended for STORM microscopy and has Ex/Em at 585/609 nm 500 uL
Catalog No. 50-004-97697 Supplier Biotium Supplier No. P005583RST500
ExoBrite STORM CF583R CD81 Antibody is designed for detection of extracellular vesicles by STORM CF583R has been validated and recommended for STORM microscopy and has Ex/Em at 585/609 nm 500 uL