Learn More
Biotium Primary Antibody CD81 P60033 CF647Plus 1/EA
Catalog No. 5000497245
Encompass_Preferred
ExoBrite STORM CF647Plus CD81 Antibody is designed for detection of extracellular vesicles by STORM CF647Plus has been validated and recommended for STORM microscopy and has Ex/Em at 652/668 nm 500 uL
Catalog No. 50-004-97245 Supplier Biotium Supplier No. P005647PST500
ExoBrite STORM CF647Plus CD81 Antibody is designed for detection of extracellular vesicles by STORM CF647Plus has been validated and recommended for STORM microscopy and has Ex/Em at 652/668 nm 500 uL