Learn More
Biotium Primary Antibody CD81 P60033 ExoBrite 640 660 1/EA
Supplier:  Biotium P005640500
Encompass_Preferred
ExoBrite 640/660 CD81 Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 640/660 nm Recommended for instruments optimized for small particle flow 100 tests
Catalog No. 50-004-97510