Learn More
Biotium Primary Antibody CD81 P60033 ExoBrite 640 660 1/EA
Catalog No. 5000499028
Encompass_Preferred
ExoBrite 640/660 CD81 Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 640/660 nm Recommended for instruments optimized for small particle flow 25 tests
Catalog No. 50-004-99028 Supplier Biotium Supplier No. P005640125
ExoBrite 640/660 CD81 Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 640/660 nm Recommended for instruments optimized for small particle flow 25 tests