Learn More
Biotium Primary Antibody CD81 P60033 ExoBrite APC 1/EA
Catalog No. 5000497937
Encompass_Preferred
ExoBrite APC CD81 Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry Recommended for instruments optimized for small particle flow 25 tests
Catalog No. 50-004-97937 Supplier Biotium Supplier No. P005APC125
ExoBrite APC CD81 Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry Recommended for instruments optimized for small particle flow 25 tests