Learn More
Biotium Primary Antibody CD8a SK1 CF503R 1/EA
Catalog No. 5000496745
Encompass_Preferred
CD8a (clone SK1) is a mouse monoclonal antibody that recognizes the human protein CD8 CD8a is a common marker of cytotoxic (Killer) T cells and is a T cell coreceptor This CF503R antibody conjugate has been validated in flow cytometry
Catalog No. 50-004-96745 Supplier Biotium Supplier No. P009503R500
CD8a (clone SK1) is a mouse monoclonal antibody that recognizes the human protein CD8 CD8a is a common marker of cytotoxic (Killer) T cells and is a T cell coreceptor This CF503R antibody conjugate has been validated in flow cytometry