Learn More
Biotium Primary Antibody CD8a SK1 CF575 1/EA
Catalog No. 5000496756
Encompass_Preferred
CD8a (clone SK1) is a mouse monoclonal antibody that recognizes the human protein CD8 CD8a is a common marker of cytotoxic (Killer) T cells and is a T cell coreceptor This CF575 antibody conjugate has been validated in flow cytometry
Catalog No. 50-004-96756 Supplier Biotium Supplier No. P009575500
CD8a (clone SK1) is a mouse monoclonal antibody that recognizes the human protein CD8 CD8a is a common marker of cytotoxic (Killer) T cells and is a T cell coreceptor This CF575 antibody conjugate has been validated in flow cytometry