Learn More
Biotium Primary Antibody CD8a SK1 PerCP 1/EA
Catalog No. 5000497700
Encompass_Preferred
CD8a (clone SK1) is a mouse monoclonal antibody that recognizes the human protein CD8 CD8a is a common marker of cytotoxic (Killer) T cells and is a T cell coreceptor This PerCP antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-97700 Supplier Biotium Supplier No. P009PCP125
CD8a (clone SK1) is a mouse monoclonal antibody that recognizes the human protein CD8 CD8a is a common marker of cytotoxic (Killer) T cells and is a T cell coreceptor This PerCP antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests