Learn More
Biotium Primary Antibody CD9 2310.9 CF405M 1/EA
Catalog No. 5000497178
Encompass_Preferred
CD9 (2310 9) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD9 This CF405M antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-97178 Supplier Biotium Supplier No. P015405M125
Add to Cart
Add to Cart
CD9 (2310 9) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD9 This CF405M antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests