Learn More
Biotium Primary Antibody CD9 2310.9 RPE Astral616 1/EA
Catalog No. 5000499109
Encompass_Preferred
CD9 (2310 9) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD9 This RPE-Astral616 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-99109 Supplier Biotium Supplier No. P015R616500
CD9 (2310 9) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD9 This RPE-Astral616 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests