Learn More
Biotium Primary Antibody CD9 24MS04.9 CF568 1/EA
Catalog No. 5000497604
Encompass_Preferred
CD9 (24MS04 9) is a recombinant rat monoclonal antibody that recognizes the mouse tetraspanin protein CD9 This CF568 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-97604 Supplier Biotium Supplier No. P016568500
Add to Cart
Add to Cart
CD9 (24MS04 9) is a recombinant rat monoclonal antibody that recognizes the mouse tetraspanin protein CD9 This CF568 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests