Learn More
Biotium Primary Antibody CD9 24MS04.9 CF647Plus 1/EA
Catalog No. 5000497605
Encompass_Preferred
CD9 (24MS04 9) is a recombinant rat monoclonal antibody that recognizes the mouse tetraspanin protein CD9 This CF647Plus antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-97605 Supplier Biotium Supplier No. P016647P125
CD9 (24MS04 9) is a recombinant rat monoclonal antibody that recognizes the mouse tetraspanin protein CD9 This CF647Plus antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests