Learn More
Biotium Primary Antibody CD9 CD63 CD81 NA ExoBrite 410 450 ExoBrite 490 515 ExoBrite 640 660 1/EA
Catalog No. 5000500447
Encompass_Preferred
ExoBrite CD9/CD63/CD81 3-Color Antibody Cocktail (Blue/Green/Far-Red) consists of three tetraspanin antibody dye conjugates in a single tube Designed for EV phenotyping by flow cytometry
Catalog No. 50-005-00447 Supplier Biotium Supplier No. P028125
ExoBrite CD9/CD63/CD81 3-Color Antibody Cocktail (Blue/Green/Far-Red) consists of three tetraspanin antibody dye conjugates in a single tube Designed for EV phenotyping by flow cytometry