Learn More
Biotium Primary Antibody CD9 CD63 CD81 NA ExoBrite 490 515 ExoBrite 560 585 ExoBrite 640 660 1/EA
Catalog No. 5000501376
Encompass_Preferred
ExoBrite CD9/CD63/CD81 3-Color Antibody Cocktail (Green/Orange/Far-Red) consists of three tetraspanin antibody dye conjugates in a single tube Designed for EV phenotyping by flow cytometry
Catalog No. 50-005-01376 Supplier Biotium Supplier No. P029500
ExoBrite CD9/CD63/CD81 3-Color Antibody Cocktail (Green/Orange/Far-Red) consists of three tetraspanin antibody dye conjugates in a single tube Designed for EV phenotyping by flow cytometry