Learn More
Biotium Primary Antibody CD9 CD63 CD81 NA ExoBrite 640 660 1/EA
Catalog No. 5000499459
Encompass_Preferred
ExoBrite CD9/CD63/CD81 Single-Color Antibody Cocktail (Far-Red) consists of three tetraspanin antibodies in a single tube all conjugated to far-red dye ExoBrite 640 Designed for high coverage EV staining for flow cytometry
Catalog No. 50-004-99459 Supplier Biotium Supplier No. P032500
ExoBrite CD9/CD63/CD81 Single-Color Antibody Cocktail (Far-Red) consists of three tetraspanin antibodies in a single tube all conjugated to far-red dye ExoBrite 640 Designed for high coverage EV staining for flow cytometry