Learn More
Biotium Primary Antibody CD9 NA CF568 1/EA
Catalog No. 5000496765
Encompass_Preferred
ExoBrite STORM CF568 CD9 (Mouse) Antibody is designed for detection of extracellular vesicles by STORM CF568 has been validated and recommended for STORM microscopy and has Ex/Em at 562/583 nm 500 uL
Catalog No. 50-004-96765 Supplier Biotium Supplier No. P018568ST500
Add to Cart
Add to Cart
ExoBrite STORM CF568 CD9 (Mouse) Antibody is designed for detection of extracellular vesicles by STORM CF568 has been validated and recommended for STORM microscopy and has Ex/Em at 562/583 nm 500 uL