Learn More
Biotium Primary Antibody CD9 NA CF647Plus 1/EA
Catalog No. 5000497538
Encompass_Preferred
ExoBrite STORM CF647Plus CD9 (Mouse) Antibody is designed for detection of extracellular vesicles by STORM CF647Plus has been validated and recommended for STORM microscopy and has Ex/Em at 652/668 nm 500 uL
Catalog No. 50-004-97538 Supplier Biotium Supplier No. P018647PST500
ExoBrite STORM CF647Plus CD9 (Mouse) Antibody is designed for detection of extracellular vesicles by STORM CF647Plus has been validated and recommended for STORM microscopy and has Ex/Em at 652/668 nm 500 uL