Learn More
Biotium Primary Antibody CD9 NA ExoBrite 560 585 1/EA
Supplier:  Biotium P018560500
Encompass_Preferred
ExoBrite 560/585 CD9 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD9 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 562/583 nm for detection in the PE channel off the yellow laser line Recommended for instruments optimized for small particle flow 100 tests
Catalog No. 50-004-97615