Learn More
Biotium Primary Antibody CD9 NA ExoBrite APC 1/EA
Catalog No. 5000497357
Encompass_Preferred
ExoBrite APC CD9 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD9 in isolated exosomes by flow cytometry Recommended for instruments optimized for small particle flow 100 tests
Catalog No. 50-004-97357 Supplier Biotium Supplier No. P018APC500
ExoBrite APC CD9 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD9 in isolated exosomes by flow cytometry Recommended for instruments optimized for small particle flow 100 tests