Learn More
Biotium Primary Antibody CD9 P21926 CF583R 1/EA
Catalog No. 5000499200
Encompass_Preferred
ExoBrite STORM CF583R CD9 Antibody is designed for detection of extracellular vesicles by STORM CF583R has been validated and recommended for STORM microscopy and has Ex/Em at 585/609 nm 500 uL
Catalog No. 50-004-99200 Supplier Biotium Supplier No. P003583RST500
ExoBrite STORM CF583R CD9 Antibody is designed for detection of extracellular vesicles by STORM CF583R has been validated and recommended for STORM microscopy and has Ex/Em at 585/609 nm 500 uL