Learn More
Biotium Primary Antibody DNP rLO-DNP-2 CF640R 1/EA
Catalog No. 5000499738
Encompass_Preferred
DNP (rLO-DNP-2) is a recombinant rat monoclonal antibody that recognizes the dinitrophenol (DNP) hapten This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99738 Supplier Biotium Supplier No. P038640R1ML
DNP (rLO-DNP-2) is a recombinant rat monoclonal antibody that recognizes the dinitrophenol (DNP) hapten This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability