Learn More
Biotium Primary Antibody DNP rLO-DNP-2 CF680R 1/EA
Catalog No. 5000499686
Encompass_Preferred
DNP (rLO-DNP-2) is a recombinant rat monoclonal antibody that recognizes the dinitrophenol (DNP) hapten This CF680R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99686 Supplier Biotium Supplier No. P038680R1ML
DNP (rLO-DNP-2) is a recombinant rat monoclonal antibody that recognizes the dinitrophenol (DNP) hapten This CF680R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability