Learn More
Biotium Primary Antibody dsRNA rJ2 Purified BSA free 1 mg mL lyophilized 1/EA
Catalog No. 5000500779
Encompass_Preferred
Anti-dsRNA (rJ2) is a recombinant mouse monoclonal antibody that detects double-stranded RNA (dsRNA) This antibody has been validated in AN-ELISA It is supplied at 1 mg/mL BSA-free and azide-free
Catalog No. 50-005-00779 Supplier Biotium Supplier No. P025500UG
Anti-dsRNA (rJ2) is a recombinant mouse monoclonal antibody that detects double-stranded RNA (dsRNA) This antibody has been validated in AN-ELISA It is supplied at 1 mg/mL BSA-free and azide-free