Learn More
Biotium Primary Antibody GFAP rGA5 CF594 1/EA
Catalog No. 5000501322
Encompass_Preferred
GFAP (rGA5) is a recombinant mouse monoclonal antibody that recognizes GFAP This CF594 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-01322 Supplier Biotium Supplier No. P0465941ML
Add to Cart
Add to Cart
GFAP (rGA5) is a recombinant mouse monoclonal antibody that recognizes GFAP This CF594 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability