Learn More
Biotium Primary Antibody GFAP rGA5 CF640R 1/EA
Catalog No. 5000501323
Encompass_Preferred
GFAP (rGA5) is a recombinant mouse monoclonal antibody that recognizes GFAP This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-01323 Supplier Biotium Supplier No. P046640R1ML
Add to Cart
Add to Cart
GFAP (rGA5) is a recombinant mouse monoclonal antibody that recognizes GFAP This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability