Learn More
Biotium Primary Antibody GFAP rGA5 CF647 1/EA
Catalog No. 5000499688
Encompass_Preferred
GFAP (rGA5) is a recombinant mouse monoclonal antibody that recognizes GFAP This CF647 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99688 Supplier Biotium Supplier No. P0466471ML
GFAP (rGA5) is a recombinant mouse monoclonal antibody that recognizes GFAP This CF647 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability